If a green color appears on the membrane after the transfer using the iBlot 2 Dry Blotting System, will it affect the results?
The green discoloration is copper deposits from the transfer stack, and it does not affect the results. To minimize this effect, shake excess water off the filter paper and buffer from the gel before placing each on the stack. The current formulation of stacks minimizes the green discoloration.
Can I cut the bottom stack of my iBlot 2 Transfer Stack to fit my separating gel?
No. Do not trim the iBlot 2 Transfer Stacks to fit your gel size.
Is the PVDF membrane in the iBlot 2 PVDF Gel Transfer Stack pre-activated?
The PVDF membrane is pre-activated and ready for use without any pre-treatment with alcohol.
Can I use the stack and/or absorbent pad more than once when using the iBlot 2 Dry Blotting System?
No. The transfer stacks have a finite amount of ions to drive the transfer and are depleted after a single use.
Can I perform multiple transfers, with a new transfer starting immediately after the previous one when using the iBlot 2 Dry Blotting System?
Yes. The device is designed to do so with no impact on performance.
Can I create my own transfer conditions with the iBlot 2 Dry Blotting System?
Yes. The device allows programming custom methods.
What types of membranes are available in the iBlot Transfer Stacks?
iBlot Transfer Stacks are available with either an integrated 0.2 µm nitrocellulose membrane or a 0.2 µm PVDF membrane. Both types are available in two sizes: a mini size which can accommodate one mini gel (8 x 8 cm), or regular size which can accommodate one E-PAGE gel, one midi gel (8 x 13 cm), or two mini gels (8 x 8 cm).
The catalog numbers are as follows:
IB3010-01 iBlot Gel Transfer Stacks, Regular (10 Blots)
IB3010-02 iBlot Gel Transfer Stacks, Mini (10 Blots)
IB4010-01 iBlot Transfer Stack PVDF, Regular (10 Blots)
IB4010-02 iBlot Transfer Stack PVDF, Mini (10 Blots)
If you would like to blot proteins onto your own membrane rather than the supplied nitrocellulose or PVDF, you can replace the integrated membrane with your desired membrane as described below:
1) Wet the desired blotting membrane in deionized water. (For PVDF membranes, first wet the PVDF membrane in 100% methanol and then rinse in deionized water).
2) Carefully remove the nitrocellulose membrane from the bottom stack using forceps.
3) Place the wetted blotting membrane on the bottom transfer stack. (Membrane should be completely wet, but not dripping; make sure there is not too much excess water.)
4) Be sure to align the membrane flush to the bottom stack gel, and remove any air bubbles using the blotting roller. Proceed with the standard iBlot transfer protocol.
Are the iBlot 3 Transfer Stacks compatible with the original iBlot and iBlot 2 transfer devices?
Unfortunately, the iBlot 3 Transfer Stacks are not compatible with the original iBlot and iBlot 2 transfer devices. The iBlot 3 Transfer Stacks are only compatible with the iBlot 3 Western Blot Transfer Device.
How many gels can I transfer simultaneously using iBlot 2 Transfer Stacks?
You can transfer:
- One midi gel or two mini gels (head-to-head) using the iBlot 2 Regular Transfer Stacks
- One mini gel using iBlot 2 Mini Transfer Stacks
I used the iBlot 2 Dry Blotting System and noticed that the signal intensity was similar for different protein loads after detection. Why is this?
This is most likely due to the protein load being too high, as a result of which detection is not within the linear range. Since the immunodetection sensitivity is higher for dry blotting with the iBlot 2 Gel Transfer Device than for semi-dry or wet blotting, we recommend that you decrease the protein load, use more diluted antibody, or perform detection for shorter time. You may need to perform some optimization based on your initial results.
I used the iBlot 2 Dry Blotting System and got high background on the membrane. Can you offer some tips?
This is likely due to use of TBST buffers for washing. We recommend using PBST or WesternBreeze wash solutions.
I used PVDF Gel Transfer Stacks with the iBlot 2 Dry Blotting System and my proteins did not transfer to the membrane. What happened?
This is likely due to the PVDF membrane being dry or partially dry. Regions where PVDF membranes are dry appear whiter than places where the membrane is wet. Remove the membrane and reactivate in 100% methanol, and rinse in water before reapplying to the transfer stack.
I transferred my proteins using the iBlot 2 Dry Blotting System and my protein bands are distorted on the membrane. Why is this?
This indicates that a non-uniform electric field was created around the wells. Ensure that the gel is properly flattened by using the Blotting Roller. Follow the recommendations on page 19 in the manual (http://tools.thermofisher.com/content/sfs/manuals/iblot2_device_man.pdf) to obtain good results.
With the iBlot 2 Dry Blotting System, some of my proteins blew through the membrane. What could have caused this?
This could happen if the transfer time used is too long. We recommend reducing the transfer time by 30 second increments.
Note: Pre-stained markers are charged, so tend to blow-through more than regular proteins.
I transferred my proteins using the iBlot 2 Dry Blotting System but the high-molecular weight proteins remained in the gel indicated by staining of the gel after transfer. Can you help me troubleshoot?
This could happen if an incorrect voltage Method was used or if inappropriate transfer conditions were used. Make sure that the voltage Method and run time used is correct, based on the gel type, as described on page 17 in the manual (http://tools.thermofisher.com/content/sfs/manuals/iblot2_device_man.pdf).
For mini or midi gels:
- Perform an ethanol equilibration step as described on page 35 in the manual (http://tools.thermofisher.com/content/sfs/manuals/iblot2_device_man.pdf) to improve transfer.
- Use a lower gel percentage to separate the high-molecular weight proteins.
- Increase the transfer time in 30-second increments.
For E-PAGE gels:
- Increase the transfer time in 30-second increments.
- Use Method P3 for 8 minutes.
Note: It is normal for some proteins to remain in the gel because some high-molecular weight proteins do not transfer completely using the iBlot 2 Gel Transfer Device, compared to semi-wet transfer apparatus.
I see a lot of empty spots on the membrane after transfer using the iBlot 2 Dry Blotting System. What could have caused this?
Here are possible causes and solutions:
- Presence of air bubbles between the gel and the membrane preventing the transfer of proteins. Be sure to remove all air bubbles between the gel and membrane using the Blotting Roller.
- Expired or creased membranes used. Use the iBlot 2 Transfer Stacks before the expiration date printed on the package.
I used the iBlot 2 Dry Blotting System and none of my proteins transferred to the membrane. What could have happened?
This could be due to no current passing through or because an incorrect voltage Method was used. Make sure that the electrical circuit is complete and current is flowing through the device. Please check to make sure that the correct voltage Method is used, see page 17 in the manual (http://tools.thermofisher.com/content/sfs/manuals/iblot2_device_man.pdf).
After transferring using the iBlot 2 Dry Blotting System, I noticed that the Bottom Stack transfer gel melted to a viscous blue solution. Why is this?
This happens if the membrane is trimmed to fit the gel size resulting in direct contact between the Top and Bottom stacks. This can be avoided by maintaining the membrane size identical to the transfer stack. Transfer quality is not affected by smaller gel size compared to the membrane.
I used the iBlot 2 Dry Blotting System and noticed some green discoloration of the edges of the membrane. Will this affect the results?
The green discoloration is due to copper ions carried with liquids that get deposited onto the membrane. These deposits do not interfere with downstream processes. The stained regions can be cut away, but membrane washing typically results in their removal.
After transferring using the iBlot 2 Dry Blotting System, both my membrane and the gel turned blue. Is this normal?
Longer transfer times result in the deposition of copper ions. Be sure to perform the transfer for the recommended time for each gel type.
I used the iBlot 2 Dry Blotting System and noticed that the Top Stack has been corroded. What could have caused this?
This could be caused by incorrect placement of the Top Stack. Please check to make sure that the Top Stack is placed correctly with the copper electrode facing up. Avoid placing the top stack in the inverted position.
I am using the iBlot 2 Dry Blotting System and no current seems to be passing through. Can you help me troubleshoot?
This indicates an incomplete electric circuit. Here are possible causes and solutions:
- Incorrect placement of iBlot 2 Absorbent Pad contact: Make sure the electrical contact of the iBlot 2 Absorbent Pad is aligned with the corresponding electrical contacts on the blotting surface of the iBlot2 Gel Transfer Device.
- Top Stack placed on the device upside down: Make sure the Top Stack is assembled with the copper electrode facing up.
- The metal safety contacts in the lid hinge may be dirty and do not make contact: Clean the metal safety contacts in the lid hinge with a cotton swab and water.
- Plastic separator was not removed when assembling stack: Make sure that the plastic separator is removed from the stack, see page 21 in the manual (http://tools.thermofisher.com/content/sfs/manuals/iblot2_device_man.pdf).
I am transferring a Tris-Glycine gel using constant voltage and the current reading is way over the expected starting current. Can you offer some suggestions?
The most common cause of abnormally high current is the transfer buffer. If the transfer buffer is too concentrated, this leads to increased conductivity and current. High current may also occur if Tris-HCl is accidentally substituted for the Tris base required in the transfer buffer. This will again result in low buffer pH and lead to increased conductivity and current and subsequently, overheating. We recommend checking the transfer buffer and its reagent components and re-diluting or remaking the buffer.
I ran my protein under native conditions on a Tris-Glycine gel. It has a pI that is higher than the pH of the Tris-Glycine transfer buffer. Can you offer some tips for transferring it?
- Increase the pH of Tris-Glycine transfer buffer to 9.2, allowing all the proteins below pI 9.2 to transfer towards the anode electrode.
- Use the Tris-Glycine transfer buffer and place a membrane on both sides of the gel. If there are any proteins that are more basic than the pH of the transfer buffer, they will be captured on the extra membrane placed on the cathode side of the gel. Both membranes can then be developed in the same manner.
- Prior to blotting, incubate the gel for 15 minutes in Tris-Glycine transfer buffer containing 0.1% SDS. The small amount of SDS will give the proteins enough charge to move unidirectionally towards the anode and in most cases, should not denature the protein. Proceed with the transfer using regular Tris-Glycine transfer buffer.
I had problems transferring my larger-molecular weight proteins from my NuPAGE gel. Can you please offer some suggestions?
For proteins larger than 100 kDa, we recommend pre-equilibrating the gel in 2X NuPAGE Transfer buffer (without methanol) containing 0.02-0.04% SDS for 10 minutes before assembling the sandwich and then transferring using 1X NuPAGE transfer buffer containing methanol and 0.01% SDS.
What causes empty spots on my membrane after transfer?
Here are possible causes and solutions:
- Presence of air bubbles between the gel and the membrane preventing the transfer of proteins. Be sure to remove all air bubbles between the gel and membrane by rolling a glass pipette over the membrane surface.
- Expired or creased membranes used. Use fresh, undamaged membranes.
I performed a western transfer and see the appearance of diffuse bands and swirling patterns on the membrane. What could have happened?
The swirling and diffuse banding patterns are typical of molecules moving laterally before binding to the membrane during transfer. Here are possible causes and solutions:
- Poor contact between the gel and the membrane: The gel should be attached to the membrane through capillary action. To ensure that this happens, make sure that you roll over the surface of each layer of the gel/membrane sandwich with a glass pipette to ensure good contact between the gel and the membrane. It is helpful to use a disposable pipette to place some extra transfer buffer on the surface of each layer as the sandwich is being made. Also, the pads need to be fully saturated (push down with gloved hand when they are placed in transfer buffer to make sure there are no air bubbles.)
- Under-compression of the gel: The gel/membrane assembly should be held securely between the two halves of the blot module. Try adding another pad or replace any pads that have lost their resiliency with fresh ones.
- Over-compression of the gel: A good indication of over-compression is if the gel has been excessively flattened. In the event that the sandwich is over-compressed, remove enough pads so that the blotter can be closed without exerting excess pressure on the gel and membrane.
Note: The height of the uncompressed pads should be 0.5-1.0 cm above the level of the sealing gasket.
When I perform a western transfer, the power supply shuts off in the middle of the transfer. What is wrong?
Here are possible causes and solutions:
- High ionic strength of the transfer buffer. Prepare the buffer as described in the manual.
- Power supply is operating at a current close to the current limit of the power supply. Use a power supply with higher limits.
After a western transfer, I noticed that a significant amount of protein remained in the gel indicated by staining of the gel after transfer. What should I do?
Here are possible causes and solutions:
- Too short a transfer time: Increase the blotting time by 15 minute increments.
- Inappropriate gel type: Check the percentage of the gel used and switch to a higher percentage gel.
- Inappropriate amount of SDS: Add 0.01-0.02% SDS to the transfer buffer to facilitate migration of the protein out of the gel.
- Inappropriate methanol content: Decrease the amount of methanol in the transfer buffer.
Note: Higher molecular weight proteins usually do not transfer completely as compared to mid to low molecular weight proteins.
After a western transfer, I noticed that a significant amount of protein had passed through the membrane indicated by the presence of proteins on the second membrane. Can you please help?
Here are possible causes and solutions:
- Too long a transfer tim: Shorten the transfer time by 15 minute increments.
- Inappropriate amount of SDS: Do not include any SDS in the transfer buffer.
- Inappropriate methanol content: Add additional methanol to the transfer buffer to increase the binding capacity of the membrane.
- Inappropriate gel type: Check the percentage of the gel used and switch to a higher percentage gel.
- Sample overloaded: Decrease the sample load.
- Finally, if using nitrocellulose membrane, switch to PVDF which has a higher binding capacity.
I performed a western transfer and none of my proteins transferred to the membrane. Can you offer some tips?
It is possible that the gel/membrane sandwich was assembled in the reverse direction such that the proteins have migrated out into the buffer. Assemble the blot sandwich in the correct order using instructions provided in the manual.
During western transfer conditions using constant voltage, what would cause the actual current to greatly exceed the expected starting current?
The most common cause of abnormally high current is the buffer. If the buffer is too concentrated, this leads to increased conductivity and higher current. High current may also occur if Tris-HCl was accidentally substituted for the Tris base required in the transfer buffer. Tris-HCl results in a low buffer pH and leads to increased conductivity and current, and, subsequently, overheating. Check the transfer buffer and its reagent components, re-dilute, or remake the buffer.
I am transferring my gel using constant voltage and the current reading has dropped much lower than the expected starting current. What could have happened?
Here are possible causes and solutions:
- The buffer was accidentally made too dilute, therefore increasing resistance and thus lowering conductivity and current: Check the transfer buffer and its reagent components and then re-dilute it or remake it.
- The circuit is broken or impeded, as in the case of a corroded or broken electrode or malfunctioning power supply: Check the equipment.
- There is a leak in the blot module (this is indicated by a drastic decrease in current and in buffer volume within the module): Ensure that the inner buffer chamber is filled sufficiently so that the wells are covered with buffer.
- Tape at the bottom of the gel cassette was not removed: Double check that the tape on the bottom of the gel has been removed.
What is the iBlot 2 Absorbent Pad?
The iBlot 2 Absorbent Pad absorbs any excess liquid formed on the stacks during blotting and generates even pressure on the stack assembly. It is placed on top of the assembled iBlot 2 stack prior to transfer. We recommend discarding the iBlot 2 Absorbent Pad after every use.
What is the Blotting Roller that is provided with the iBlot 2 Dry Blotting System?
The Blotting Roller is a plastic roller attached to a stainless steel handle. It is used to remove any air bubbles between the gel and blotting membrane during the assembly of the stacks and gel.
Can I cut the membrane in the iBlot 2 Transfer Stacks to fit my separating gel?
No. Maintain the membrane size identical to the transfer stacks. This helps ensure that there is no direct contact between the top and bottom transfer stacks.
Are the iBlot 2 PVDF and nitrocellulose membranes compatible with fluorescent antibodies or membrane staining (e.g., LI-COR, Ponceau S)?
The membranes are compatible with all commonly used detection methods such as staining, immunodetection, fluorescence, etc.
Can I replace the membrane with my preferred membrane when using the iBlot 2 Dry Blotting System?
It is possible but the results may be inferior.
How can I get better transfer of high-molecular weight proteins using the iBlot 2 Dry Blotting System?
Proteins larger than ~150 kDa migrate more slowly than smaller proteins. Therefore, more time is required to transfer them from the gel to the membrane. We recommend using a transfer time of 8-10 minutes for optimal transfer of these proteins.
To enhance transfer efficiency, we also recommend adding an equilibration (gel-soaking) step between electrophoresis and western transfer and using NuPAGE 3-8% Tris-acetate gels for electrophoresis. For the protocol, please refer to page 35 in the manual (http://tools.thermofisher.com/content/sfs/manuals/iblot2_device_man.pdf).
Can the iBlot 2 Dry Blotting System be used for transfer of low- and high-molecular weight proteins?
Yes. However, minor optimizations in the transfer protocol (i.e., steps, voltage, time) may be needed for some proteins.
Are the copper electrodes in the transfer stacks of the iBlot 2 Dry Blotting System recyclable?
To maximize the recovery of the copper mesh electrodes used in our iBlot 2 transfer stacks, we have established an agreement with a recycling center in the United States. In order to prepare the copper electrodes for recycling, please follow the instructions listed here (https://www.thermofisher.com/content/dam/LifeTech/migration/files/proteins-expression-isolation-analysis/pdfs.par.0358.file.dat/iBlot%202%20copper%20reycling%20instructions%20NA%20only.pdf).
I want to perform western transfer with mini gels (8 x 8 cm), but I do not have iBlot 2 Transfer Stacks (Mini). Can I use iBlot 2 Transfer Stacks (Regular) to transfer my mini gels?
It is best not to transfer a single mini gel on a regular-sized transfer stack. Although in most cases the transfer will be fine, empty spaces on the transfer stack that are not in direct contact with a gel could potentially cause distortions across the whole surface of the membrane, including the portion in contact with the gel. It is best to have more than 50% of a membrane in contact with the gel, if possible. So the recommendation would be to transfer 2 mini gels using a Regular stack.
Unlike the iBlot Gel Transfer Stacks, in the iBlot 2 Top Stack, I see a white sheet beneath the Transfer gel layer. What purpose does it serve?
This is the plastic separator that separates the Top Stack from the Bottom Stack. Right before assembling the stack, this separator needs to be removed from the Top Stack and discarded.
Note: In some instances, the membrane may adhere to the separator. If this is the case, use forceps to remove the membrane and place it on top of the Bottom Stack.
Do you offer replacement electrodes for the iBlot 2 Gel Transfer Device?
Yes, we offer the iBlot 2 Electrode Replacement Kit, Cat. No. IB28001, that consists of two electrical contacts, two screws, and a round bumper. Please refer to page 37 in the manual (http://tools.thermofisher.com/content/sfs/manuals/iblot2_device_man.pdf) for the instructions.
Is it okay to freeze the iBlot 2 Gel Transfer Stacks?
The iBlot 2 Gel Transfer stacks should not be frozen as they contain a gel matrix that will be damaged by freezing.
Can I use the iBlot 2 Transfer Stacks after the expiration date?
No. The results may be inferior after the expiration date.
What is the shelf life of iBlot 2 Gel Transfer Stacks?
It is currently 9 months, and the expiration date is printed on the box.
What is the recommended storage condition for the iBlot 2 Transfer Stacks?
We recommend storing the iBlot 2 Transfer Stacks at room temperature.
What are the different kinds of iBlot 2 Transfer Stacks that you offer and are they available separately?
We offer iBlot 2 Gel Transfer Stacks with integrated nitrocellulose or PVDF membranes for transferring proteins from gels onto nitrocellulose or PVDF membranes, respectively (western blotting). These stacks can also be purchased separately.
What kind of gels can I use with the iBlot 2 Dry Blotting System for western blotting?
The iBlot 2 Dry Blotting System is compatible for use with Bolt Bis-Tris, NuPAGE Bis-Tris, Tris-Acetate, Tris-Glycine, Tricine (in mini- and midi-gel formats), and E-PAGE gels (E-PAGE 48 only).
What is the purpose of the iBlot Filter Paper?
The iBlot Filter Paper is used for blotting mini- or midi-gels. It is placed on top of the gel before placing the iBlot Cathode Stack (Top Stack) to protect the gel integrity during the blotting process. The iBlot Filter Paper is supplied in two sizes for efficient blotting of mini- and midi-gels. We do not recommend using the iBlot Filter Paper for blotting E-PAGE gels.
Note: Failure to use the iBlot Filter Paper during blotting of mini- or midi-gels may result in high currents exceeding the current limit leading to an error (Error2) during the run.
Do you still offer the iBlot Gel Transfer Device?
The iBlot Gel Transfer Device has been discontinued and we have launched the iBlot 2 Gel Transfer Device, that is an improved version. The iBlot Transfer Stacks, Western Detection Stacks and DNA Transfer Stacks are still available for purchase and are to be used exclusively with the original iBlot Gel Transfer Device, and are not compatible with the new iBlot 2 Gel Transfer Device (http://www.thermofisher.com/order/catalog/product/IB21001). Only iBlot 2 consumables are to be used with the iBlot 2 Gel Transfer Device.
Can I use the iBlot Filter Paper for blotting E-PAGE gels?
We do not recommend using the iBlot Filter Paper for blotting E-PAGE gels.
What method do you recommend for blotting E-PAGE gels?
The preferred method of transfer for E-PAGE gels is dry blotting using the iBlot/iBlot 2 Dry Blotting System (see Page 27 of the E-PAGE Technical Guide http://tools.thermofisher.com/content/sfs/manuals/epagetechguide_man.pdf). Please note that this works only for E-PAGE 48 gels. Semi-dry blotting using the Invitrogen Semi-Dry Blotter, Cat. No. SD1000 (Page 33), or semi-wet blotting (Page 37) may also be performed.
Does the iBlot Dry Blotting System work with native or native-blue gels?
Yes, please take a look at the Western Blotting NativePAGE Invitrogen Bis-Tris Gels Using the iBlot 7-Minute Blotting System Application Note (http://www.thermofisher.com/content/dam/LifeTech/migration/en/filelibrary/pdf.par.18870.file.dat/native-with-iblot-app-note-v3.pdf).