Learn More
Invitrogen™ Coverglass Removal Tool DFS Item
Minimize the risk of breakage of chambered coverglass during cell culture and fluorescence imaging applications. CultureWell&Trade; Coverglass Removal Tool enables the chambered upper structure to be easily separated from the coverglass without the need for excessive force.
Supplier:  Invitrogen™ C37010
Catalog No. C37010
Only null left
Description
Description
The Coverglass Removal Tool is intended for use with a CultureWell™ Chambered Coverglass for Cell Culture (Mfc. Nos. C37000, C37005). The removable chambered coverglass is provided sterile and ready to use. It contains 16 wells that can each hold up to 250μL, allowing cells to be cultured in a number of different conditions on a single slide. When the cells are ready to be imaged, coverglass removal is made easy by the use of this simple tool.
Specifications
Specifications
Coverglass Removal Tool
Culturewell
Microscope
1 each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.