missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Countess™ USB Drive

Catalog No. A25751
Encompass
Change view
Click to view available options
Quantity:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25751 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A25751 Supplier Invitrogen™ Supplier No. A25751

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Accessory for Countess II and Countess II FL Automated Cell Counter

For use with Countess® II & Countess® II FL automated cell counters, as well as other USB-enabled devices.

Specifications

Type USB Drive
Product Line Countess
For Use With (Application) Cell Counting, Viability, Cryopreservation
For Use With (Equipment) Countess™ Automated Cell Counter
Quantity 1 unit
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.