missing translation for 'onlineSavingsMsg'
Learn More

Basco™ Life Latch™ New Generation Bucket with Screw Top Lid, 5 gal.

Catalog No. 19339067
Change view
Click to view available options
Capacity (English):
5 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19339067 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19339067 Supplier Basco™ Supplier No. SS050WH

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Keep your products secured and contained in this FDA-compliant bucket with screw on lid.

Keep products secure with this 5 gallon bucket with screw on lid and a unique trigger mechanism to automatically secure the lid when closing.

  • Built-in plastic handle for convenient carrying
  • Removable and reusable plastic cover
  • FDA-compliant food storage container
  • Easy finger activated release for easy opening of lid
  • UN Rated screw top plastic containers for hazardous materials

Specifications

Closure Included Yes
Closure Type Lid
Capacity (English) 5 gal.
Capacity (Metric) 19 L
Color White
Material HDPE
Shape Round
Height (English) 11.5 in.
Diameter (English) Opening 11.25 in.
Weight (English) 2.36 lb.