missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Trimethylphenylammonium hydroxide Solution, ∼0.5 M (CH3)3N(OH)C6H5 in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111018457
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018457 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018457 Supplier Merck Emd Millipore Supplier No. 7926650ML

Trimethylphenylammonium hydroxide (TMAH) is a methylating reagent.

Chemical Identifiers

MDL Number MFCD00041899
Synonym Phenyltrimethylammonium hydroxide; TMAH

Specifications

CAS 1899-02-1
Linear Formula (CH3)3 N(OH)C6H5
Quantity 50 mL
Formula Weight 153.22
Grade For GC derivatization
Concentration or Composition (by Analyte or Components) 0.5 M (CH3)3 N(OH)C6H5 in. methanol
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany