missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Trifluoroacetic Anhydride, For GC derivatization, 99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7193493
Change view
Click to view available options
Quantity:
10 mL
50 mL
10 x 1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018735 10 x 1 mL
111019765 10 mL
111018363 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111018735 Supplier Merck Emd Millipore Supplier No. 9171910X1ML
Only null left

Reagent for cabethoxy, butyl ester, n-methyl, methyl ester, di(trifluoroacetyl), hexafluoroisopropyl ester, acetyl or trifluoroacetyl phenylthihydantoins, trifluoroacetyl (2-hydroxy-2-methyl)ethyl, n-trifluoroacetyl, trifluoroacetyl (TFA), trifluoroacetamide, 4-trifluoromethyl-2, 6-diamino-1,3,5-triazineacetyl and trifluoroacetyl vanillin.

Chemical Identifiers

CAS 407-25-0
Molecular Formula C4F6O3
Molecular Weight (g/mol) 210.03
MDL Number MFCD00000416
InChI Key QAEDZJGFFMLHHQ-UHFFFAOYSA-N
Synonym TFAA
IUPAC Name 2,2,2-trifluoroacetyl 2,2,2-trifluoroacetate
SMILES FC(F)(F)C(=O)OC(=O)C(F)(F)F

Specifications

Melting Point -65°C
Boiling Point 39.5°C to 40°C
Linear Formula (CF3CO)2 O
Formula Weight 210.03
Percent Purity ≥99.0% (GC)
Grade GC/MS (SIM)
Density 1.511 g/mL (at 20°C)
Quantity 10 x 1 mL
Refractive Index n20/D 1.3
Vapor Pressure 6.28 psi (20°C)
RTECSNumber AJ9800000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany