missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Tetramethylammonium sulfate , ≥99.0% (T), MilliporeSigma™ Supelco™

Catalog No. 111013086
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013086 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013086 Supplier Merck Emd Millipore Supplier No. 0279910G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00012139
Synonym Bis(tetramethylammonium) sulfate

Specifications

CAS 14190-16-0
Linear Formula [(CH3)4 N]2(SO4)
Quantity 10 g
Formula Weight 244.35
Percent Purity ≥99.0% (T)
Grade For ion pair chromatography
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany