missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Tetramethylammonium bromide , ≥99.0% (AT), MilliporeSigma™ Supelco™

Catalog No. 111018249
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018249 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018249 Supplier Merck Emd Millipore Supplier No. 8770810G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00011626

Specifications

Melting Point >300°C
CAS 64-20-0
Linear Formula (CH3)4 N(Br)
Quantity 10 g
Formula Weight 154.05
Percent Purity ≥99.0% (AT)
Grade For ion pair chromatography
Product Line LiChropur
RTECSNumber BS7600000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany