missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ Tetrakis(decyl)ammonium bromide , ≥99.0% (AT), MilliporeSigma™ Supelco™
Specifications
Specifications
| Melting Point | 87°C to 90°C |
| CAS | 14937-42-9 |
| Physical Form | Powder |
| Quantity | 50 g |
| Linear Formula | [CH3(CH2)9]4 N(Br) |
| Molecular Weight (g/mol) | 659.01 |
| Formula Weight | 659.01 |
| Percent Purity | ≥99.0% (AT) |
| Grade | Ion Pair Chromatography |
| Product Line | LiChropur |
| Show More |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany