missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Tetrakis(decyl)ammonium bromide , ≥99.0% (AT), MilliporeSigma™ Supelco™

Catalog No. 111013183
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013183 10 g
111013184 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013183 Supplier Merck Emd Millipore Supplier No. 8757810GF

Chemical Identifiers

MDL Number MFCD00043166
Synonym Tetra-decylammonium bromide

Specifications

Melting Point 87°C to 90°C
CAS 14937-42-9
Quantity 10 g
Linear Formula [CH3(CH2)9]4 N(Br)
Formula Weight 659.01
Percent Purity ≥99.0% (AT)
Grade Ion Pair Chromatography
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany