missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Tetraethylammonium hydrogen sulfate,, For ion pair chromatography, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111013325
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013325 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013325 Supplier Merck Emd Millipore Supplier No. 8662610GF

Chemical Identifiers

MDL Number MFCD00036150

Specifications

Melting Point 243°C to 245°C
CAS 16873-13-5
Linear Formula (C2H5)4 N(HSO4)
Quantity 10 g
Formula Weight 227.32
Percent Purity ≥99.0% (T); ≥99.0%
Grade For ion pair chromatography
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany