missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Tetrabutylammonium phosphate monobasic solution , MilliporeSigma™ Supelco™

Catalog No. 111014609
Change view
Click to view available options
Quantity:
6 x 25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014609 6 x 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014609 Supplier Merck Emd Millipore Supplier No. 868996X1AMPF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Concentrate for 6 x 1 L ready-to-use solution. Dilute contents of one ampule (∼0.3 M) to 1 L with HPLC grade water to obtain a 0.005 M eluent solution.

Chemical Identifiers

MDL Number MFCD00064526
Synonym Tetrabutylammonium dihydrogen phosphate

Specifications

CAS 5574-97-0
Quantity 6 x 25 mL
Linear Formula (CH3 CH2 CH2 CH2)4 N[OP(OH)2 O]
Formula Weight 339.45
Grade Ion Pair Chromatography
Packaging Ampule
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany