missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Tetrabutylammonium iodide,, For ion pair chromatography, ≥99.0%, MilliporeSigma™ Supelco™

$321.24 - $321.24

Chemical Identifiers

MDL Number MFCD00011636
Synonym TBAI
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111013895
View Documents
Merck Emd Millipore
8690310GF
10 g
Each for $321.24
Only null left
 
Description

Description

Specifications

Chemical Identifiers

MFCD00011636
TBAI

Specifications

141°C to 143°C; 144°C to 146°C
10 g
MFCD00011636
369.37
Ion Pair Chromatography
311-28-4
(CH3 CH2 CH2 CH2)4 N(I)
TBAI
≥99.0% (AT); ≥99.0%
LiChropur
Videos
Safety and Handling

Safety and Handling

RTECSNumber : BS5450000

SDS
missing translation for 'documents'

missing translation for 'documents'

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany