missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Tetrabutylammonium bisulfate,, For ion pair chromatography, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111013926
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
10 x 1 g
50 g
Catalog No. Quantity
111013926 10 g
111013925 10 x 1 g
111013927 50 g
3 options

Catalog No. 111013926

Supplier: Merck Emd Millipore 8685310GF

Only null left

Tetrabutyammonium bisulfate (TBAHS) is an ion-pairing reagent. It acts as a catalyst in hydrolysis of aziridines and epoxides and the catalyst can be recycled.

Chemical Identifiers

MDL Number MFCD00011637
Synonym Tetrabutylammonium hydrogen sulfate

Specifications

Melting Point 171°C to 174°C
CAS 32503-27-8
Flash Point 175.6°C - Closed Cup
Linear Formula (CH3 CH2 CH2 CH2)4 N(HSO4)
Quantity 10 g
Formula Weight 339.53
Percent Purity ≥99.0% (T); ≥99.0%
Grade For ion pair chromatography
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany