missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ Sodium sulfate, 99.0-101.0% (T), MilliporeSigma™ Supelco™
Specifications
Specifications
| Melting Point | 884°C |
| Physical Form | Solid |
| Quantity | 250 g |
| Product Line | LiChropur |
| Density | 2.68 g/mL (at 25°C) |
| Linear Formula | Na2 SO4 |
| Solubility Information | Water: 1 M, clear, colorless |
| Formula Weight | 142.04 |
| Percent Purity | 99.0 to 101.0% (T) |
| Grade | HPLC |
| Show More |
Safety and Handling
| RTECSNumber | WE1650000 |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany