missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Sodium Perchlorate Monohydrate, MilliporeSigma™ Supelco™

Catalog No. p-7214145
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022672 50 g
111022671 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111022672 Supplier Merck Emd Millipore Supplier No. 5658150G
Only null left

For HPLC

Chemical Identifiers

CAS 7-3-7791
Molecular Formula ClH2NaO5
Molecular Weight (g/mol) 140.45
MDL Number MFCD00149164
InChI Key IXGNPUSUVRTQGW-UHFFFAOYSA-M
IUPAC Name sodium hydrate perchlorate
SMILES O.[Na+].[O-][Cl](=O)(=O)=O

Specifications

pH 6 to 8
Quantity 50 g
Linear Formula NaClO4 · H2O
Solubility Information H2O: 5%, Clear, Colorless
Formula Weight 140.46
Percent Purity 99% to 101% (T)
Grade HPLC
Hazard Category H271
RTECSNumber SC9850000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany