missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Sodium 1-pentanesulfonate monohydrate , ≥99.0% (T), MilliporeSigma™ Supelco™

Catalog No. 111013513
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013513 5 g
111013511 10 g
111013512 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111013513 Supplier Merck Emd Millipore Supplier No. 769522.5GF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium 1-pentanesulfonate monohydrate is an Ion pair reagent. It is a anionic surfactant which has opposite influence on aniline polymerization thermal effect. It also has highest and lowest enthalpy of polymerization values.

Chemical Identifiers

MDL Number MFCD00149548
Synonym 1-Pentanesulfonic acid sodium salt

Specifications

CAS 207605-40-1
Linear Formula CH3(CH2)4 SO3 Na · H2O
Quantity 5 g
Formula Weight 192.21
Percent Purity ≥99.0% (T)
Grade For ion pair chromatography
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany