missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Sodium 1-decanesulfonate ,, For ion pair chromatography, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111012972
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012972 10 g
111012973 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111012972 Supplier Merck Emd Millipore Supplier No. 3063110GF

Chemical Identifiers

MDL Number MFCD00007526
Synonym 1-Decanesulfonic acid sodium salt

Specifications

Melting Point >300°C
CAS 13419-61-9
Quantity 10 g
Linear Formula CH3(CH2)8 CH2 SO3 Na
Formula Weight 244.33
Percent Purity ≥99.0% (T); ≥99.0%
Grade Ion Pair Chromatography
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany