missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ Sodium 1-butanesulfonate, For ion pair chromatography, 99.0% (T), MilliporeSigma™ Supelco™
$284.69 - $1200.88
Chemical Identifiers
| CAS | 2386-54-1 |
|---|---|
| Molecular Formula | C4H9NaO3S |
| Molecular Weight (g/mol) | 160.16 |
| MDL Number | MFCD00007540 |
| InChI Key | XQCHMGAOAWZUPI-UHFFFAOYSA-M |
| Synonym | 1-Butanesulfonic acid sodium salt |
| IUPAC Name | sodium butane-1-sulfonate |
| SMILES | [Na+].CCCCS([O-])(=O)=O |
Chemical Identifiers
| 2386-54-1 | |
| 160.16 | |
| XQCHMGAOAWZUPI-UHFFFAOYSA-M | |
| sodium butane-1-sulfonate |
| C4H9NaO3S | |
| MFCD00007540 | |
| 1-Butanesulfonic acid sodium salt | |
| [Na+].CCCCS([O-])(=O)=O |
Specifications
| >300°C | |
| 10 g | |
| CH3(CH2)3 SO3Na | |
| 1-Butanesulfonic acid sodium salt | |
| [Na+].CCCCS([O-])(=O)=O | |
| 160.16 | |
| ≥99.0% (T) | |
| LiChropur |
| 2386-54-1 | |
| C4H9NaO3S | |
| MFCD00007540 | |
| XQCHMGAOAWZUPI-UHFFFAOYSA-M | |
| sodium butane-1-sulfonate | |
| 160.17 | |
| Ion Pair Chromatography |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany