missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Sodium 1-butanesulfonate, For ion pair chromatography, 99.0% (T), MilliporeSigma™ Supelco™

Catalog No. 111013646
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013646 10 g
111013647 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013646 Supplier Merck Emd Millipore Supplier No. 1902210GF

Chemical Identifiers

CAS 2386-54-1
Molecular Formula C4H9NaO3S
Molecular Weight (g/mol) 160.16
MDL Number MFCD00007540
InChI Key XQCHMGAOAWZUPI-UHFFFAOYSA-M
Synonym 1-Butanesulfonic acid sodium salt
IUPAC Name sodium butane-1-sulfonate
SMILES [Na+].CCCCS([O-])(=O)=O

Specifications

Melting Point >300°C
Quantity 10 g
Linear Formula CH3(CH2)3 SO3Na
Formula Weight 160.17
Percent Purity ≥99.0% (T)
Grade Ion Pair Chromatography
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany