missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Phosphoric Acid Solution, For HPLC, 49.5-50.5%, MilliporeSigma™ Supelco™

Catalog No. 111019330
Change view
Click to view available options
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111019330 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019330 Supplier Merck Emd Millipore Supplier No. 93752250ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 7664-38-2
Density 1.331 to 1.336 g/mL (at 20°C)
Quantity 250 mL
Linear Formula H3PO4
MDL Number MFCD00011340
Formula Weight 98.00
Percent Purity 49.5 to 50.5%
Grade HPLC

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany