missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Pentafluoropropionic anhydride, 99%, MilliporeSigma™ Supelco™

Catalog No. p-7187707
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
10 x 1 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019174 5 mL
111018609 10 x 1 mL
111018398 25 mL
111017965 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 111019174 Supplier Merck Emd Millipore Supplier No. 3949045ML
Only null left

Chemical Identifiers

MDL Number MFCD00000429
Synonym PFPA; Perfluoropropionic anhydride

Specifications

CAS 356-42-3
Density 1.571 g/mL (at 25°C)
Boiling Point 69°C to 70°C (735 mmHg)
Packaging Ampule
Linear Formula (CF3 CF2 CO)2 O
Quantity 5 mL
Formula Weight 310.05
Percent Purity 99%
Grade For GC derivatization
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany