missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ N-(Propionyloxy)succinimide, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013864
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013864 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013864 Supplier Merck Emd Millipore Supplier No. 935351G
Only null left

Chemical Identifiers

MDL Number MFCD03427483
Synonym 1-(1-Oxopropoxy)-2,5-pyrrolidinedione; 2,5-Dioxopyrrolidin-1-yl-propanoate; Propanoic acid-NHS ester; Propionyl-Osu; Succinimidyl propionate

Specifications

CAS 30364-55-7
Linear Formula C7H9 NO4
Quantity 1 g
Formula Weight 171.15
Percent Purity ≥95% (HPLC)
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany