missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, MilliporeSigma™ Supelco™
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane is a silylating reagent generally used in estrogenic steroid hormone identification and quantification.
Supplier: Merck Emd Millipore T638110AMP
Specifications
| 25561-30-2 | |
| 45°C to 50°C (14 mmHg) | |
| Ampule | |
| MFCD00008269 | |
| BSTFA + TMCS | |
| For GC derivatization |
| 0.97 g/mL | |
| 6°C | |
| CF3 C[=NSi(CH3)3 ]OSi(CH3)3 | |
| 10 x 1 mL | |
| 257.40 | |
| LiChropur |
Chemical Identifiers
| MFCD00008269 | |
| BSTFA + TMCS |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany