missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, 99% (excluding TMCS), MilliporeSigma™ Supelco™

Catalog No. p-7193414
Change view
Click to view available options
Quantity:
25 mL
10 x 0.1 mL
10 x 1 mL
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
111019003 10 x 0.1 mL Glass Bottle
111018800 10 x 1 mL Glass Bottle
111018739 25 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111019003 Supplier Merck Emd Millipore Supplier No. 1523810X0.1ML
Only null left

For GC Derivatization, Contains 1% TMCS

Silylating mixture used for the silylation of various compounds

Chemical Identifiers

CAS 25561-30-2
Molecular Formula C8H18F3NOSi2
Molecular Weight (g/mol) 257.40
MDL Number MFCD00008269
InChI Key XCOBLONWWXQEBS-UHFFFAOYSA-N
Synonym Silylating mixture V; BSTFA + TMCS
IUPAC Name trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)ethanimidate
SMILES C[Si](C)(C)OC(=N[Si](C)(C)C)C(F)(F)F

Specifications

Density 0.97 g/mL
Boiling Point 45°C to 50°C (14 mmHg)
Packaging Glass Bottle
Linear Formula CF3C[=NSi(CH3)3 ]OSi(CH3)3
Refractive Index n20/D 1.384
Quantity 10 x 0.1 mL
Formula Weight 257.40
Percent Purity 99% (excluding TMCS)
Grade For GC derivatization
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany