missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ N,O-Bis(trimethylsilyl)trifluoroacetamide, For GC Derivatization, 99.0%, MilliporeSigma™ Supelco™

Catalog No. 111018715
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
111018715 25 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018715 Supplier Merck Emd Millipore Supplier No. 1522225MLF

missing translation for 'validMainframeMsg'

N,O-Bis(trimethylsilyl)trifluoroacetamide (BSTFA) is an excellent silylation reagent. For some sterically hindered primary or secondary nitro compounds, N,O-bis(trimethylsilyl)acetamide and BSTFA achieve better results than normal silylation conditions. BSTFA finds application in derivatizing amines. It is suitable for silyaltion of carboxylic acids, phenols, ureas, amides and alcohols.

Chemical Identifiers

CAS 25561-30-2
Molecular Formula C8H18F3NOSi2
Molecular Weight (g/mol) 257.40
MDL Number MFCD00008269
InChI Key XCOBLONWWXQEBS-UHFFFAOYSA-N
Synonym BSTFA
IUPAC Name trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)ethanimidate
SMILES C[Si](C)(C)OC(=N[Si](C)(C)C)C(F)(F)F

Specifications

Density 0.969 g/mL (at 25°C)
Boiling Point 45°C to 50°C (14 mmHg)
Packaging Glass Bottle
Linear Formula CF3C[=NSi(CH3)3 ]OSi(CH3)3
Refractive Index n20/D 1.384; n20/D 1.384
Quantity 25 mL
Formula Weight 257.40
Percent Purity ≥99.0%
Grade For GC derivatization
Physical Form Liquid
Product Line LiChropur
Show More Show Less

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany