missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ N,O-Bis(trimethylsilyl)trifluoroacetamide, For GC derivatization, ≥99.0%, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 25561-30-2 |
| Density | 0.969 g/mL (at 25°C) |
| Boiling Point | 45°C to 50°C (14 mmHg) |
| Flash Point | 24°C |
| Packaging | Glass Bottle |
| Linear Formula | CF3 C[=NSi(CH3)3 ]OSi(CH3)3 |
| Quantity | 1 mL |
| Molecular Weight (g/mol) | 257.40 |
| Formula Weight | 257.40 |
| Percent Purity | ≥99.0% (GC); ≥99.0% |
| Show More |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany