missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ N-Methyl-N-trimethylsilyltrifluoroacetamide Activated III, For GC Derivatization, MilliporeSigma™ Supelco™
Description
Derivatizing agent for the per(trimethylsilylation) of analytes for GC and GC-MS
Specifications
Specifications
| Chemical Name or Material | N-Methyl-N-trimethylsilyltrifluoroacetamide |
| CAS | 24589-78-4 |
| Refractive Index | n20/D 1.383 |
| Quantity | 5 mL |
| MDL Number | MFCD00000411 |
| Synonym | N-Methyl-N-trimethylsilyltrifluoroacetamide imidazole mixture; MSTFA activated III |
| Grade | For GC derivatization |
| Physical Form | Liquid |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany