missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ N-Methyl-N-trimethylsilyltrifluoroacetamide Activated I, For GC Derivatization, MilliporeSigma™ Supelco™
Description
Derivatizing agent for the trimethylsilylation of -OH and -NH groups for GC and GC-MS
Specifications
Specifications
| Packaging | Clear Glass Bottle |
| Refractive Index | n20/D 1.380 |
| Quantity | 5 mL |
| Synonym | MSTFA activated I |
| Grade | For GC derivatization |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany