missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Hydrogen Chloride 2-propanol Solution, ∼1.25 M HCl (T), For GC Derivatization, MilliporeSigma™ Supelco™

Catalog No. 111017269
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017269 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017269 Supplier Merck Emd Millipore Supplier No. 17933250ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Density 0.821 g/mL (at 20°C)
Quantity 250 mL
Linear Formula HCl · CH3CH(OH)CH3
MDL Number MFCD00011324
Synonym Hydrochloric acid in 2-propanol
Formula Weight 36.46
Grade GC

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany