missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Hexadecyltrimethylammonium bisulfate, ≥99.0% (T), MilliporeSigma™ Supelco™

Catalog No. 111015157
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015157 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015157 Supplier Merck Emd Millipore Supplier No. 523715GF

Chemical Identifiers

MDL Number MFCD00134393
Synonym Hexadecyltrimethylammonium hydrogen sulfate

Specifications

CAS 68214-07-3
Packaging Glass Bottle
Linear Formula CH3(CH2)15 N(HSO4)(CH3)3
Quantity 5 g
Formula Weight 381.61
Percent Purity ≥99.0% (T)
Grade For ion pair chromatography
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany