missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Ethylenediaminetetraacetic acid disodium salt dihydrate, 99.0-101.0% (KT), MilliporeSigma™ Supelco™

Catalog No. p-7187611
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015015 10 g
111015016 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015015 Supplier Merck Emd Millipore Supplier No. 7988410G
Only null left

Chemical Identifiers

Synonym Disodium ethylenediaminetetraacetate dihydrate; EDTA disodium salt; EDTA-Na2; Edathamil; Edetate disodium salt dihydrate; Sequestrene Na2

Specifications

Melting Point 248°C
CAS 6381-92-6
Quantity 10 g
Linear Formula C10H14N2 Na2O8 · 2H2 O
Solubility Information Water: 0.1 M (at 20°C, clear, colorless)
Formula Weight 372.24
Percent Purity 99.0 to 101.0% (KT)
Grade HPLC
Product Line LiChropur
RTECSNumber AH4410000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany