missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Difluoroacetic Acid, MilliporeSigma™ Supelco™

Catalog No. p-7214122
Change view
Click to view available options
Quantity:
1 mL
50 mL
10 x 1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022365 1 mL
111022364 10 x 1 mL
111022366 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111022365 Supplier Merck Emd Millipore Supplier No. 009221ML
Only null left

Compared to trifluoroacetic acid (TFA), difluoroacetic acid (DFA) exhibits an improved sensitivity in mass spectral detection (ESI). It has a reduced effect on ion suppression in reversed-phase liquid chromatography (RPLC-MS), while keeping its strong chromatographic performance enhancing effects.

Chemical Identifiers

CAS 381-73-7
Molecular Formula C2H2F2O2
Molecular Weight (g/mol) 96.03
MDL Number MFCD00004220
InChI Key PBWZKZYHONABLN-UHFFFAOYSA-N
Synonym DFA
IUPAC Name 2,2-difluoroacetic acid
SMILES OC(=O)C(F)F

Specifications

Melting Point -1°C (literature)
Density 1.526 g/mL (at 25°C (literature))
Boiling Point 132°C to 134°C (literature)
Flash Point 78°C (Closed Cup)
Linear Formula F2CHCO2H
Refractive Index n20/D 1.344 (literature)
Quantity 1 mL
Formula Weight 96.03
Percent Purity 97.5% to 102.5% (T)
Hazard Category H314
RTECSNumber AG9900000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany