missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Dichlorodimethylsilane Solution, ∼5 in Toluene, For GC Derivatization, MilliporeSigma™ Supelco™

Catalog No. 111019238
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019238 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019238 Supplier Merck Emd Millipore Supplier No. 08471500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For deactivating glassware.Treat the glass tubing that you use for columns with Sylon-CT solution (5% dimethyldichlorosilane in toluene). It deactivates the tubing for use up to 350 C to 400 C, far exceeding the capabilities of other deactivating agents used at room temperature. Sylon CT™ also deactivates transfer lines and glass or glass-lined injection port liners. Instructions included.

Specifications

CAS 75-78-5
Quantity 500 mL
Linear Formula C2H6Cl2Si
MDL Number MFCD00000491
Synonym Sylon(TM) CT; Dimethyldichlorosilane solution
Formula Weight 129.06
Grade GC Derivatization
Product Line Sylon
Refractive Index n20/D 1.491-1.495

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany