missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ Dansyl chloride, ≥99.0% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| Melting Point | 72°C to 74°C |
| CAS | 605-65-2 |
| Quantity | 100 mg |
| Linear Formula | C12H12 ClNO2 S |
| Solubility Information | Acetone: 0.2 g/10 mL, clear to slightly turbid, yellow to very dark yellow-orange |
| Formula Weight | 269.75 |
| Percent Purity | ≥99.0% (HPLC) |
| Grade | HPLC |
| Product Line | LiChropur |
Safety and Handling
| RTECSNumber | QK3688000 |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany