missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Chlorotrimethylsilane, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020596
Change view
Click to view available options
Quantity:
10 mL
10 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020596 10 mL
111019785 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020596 Supplier Merck Emd Millipore Supplier No. 8959510ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chlorotrimethylsilane is a silylation reagent used in the derivatization of triglycerides into volatile fatty esters by gas chromatography (GC).

Chemical Identifiers

MDL Number MFCD00000502
Synonym TMCS; Trimethylchlorosilane; Trimethylsilyl chloride

Specifications

Boiling Point 57°C
Melting Point -40°C
CAS 75-77-4
Quantity 10 mL
Linear Formula (CH3)3 SiCl
Flash Point -28°C - Closed Cup
Formula Weight 108.64
Percent Purity ≥99.0% (GC)
Grade GC Derivatization
Ignition Point 752°F
Product Line LiChropur
Vapor Pressure 100 mmHg (25°C)
Density 0.856 g/mL (at 25°C); Vapor Density: 3.7 (vs. air)
Show More Show Less
RTECSNumber VV2710000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany