missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Chlorotriethylsilane, ≥97.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018187
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018187 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018187 Supplier Merck Emd Millipore Supplier No. 9038350ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00000507
Synonym TESCl; Triethylchlorosilane

Specifications

Boiling Point 142°C to 144°C
CAS 994-30-9
Quantity 50 mL
Linear Formula (C2H5)3 SiCl
Flash Point 30°C
Formula Weight 150.72
Percent Purity ≥97.0% (GC)
Grade GC Derivatization
Product Line LiChropur
Density 0.898 g/mL (at 25°C)

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany