missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Boron Trifluoride - 1-butanol Solution, ∼10% in 1-butanol (1.3 M), For GC Derivatization, MilliporeSigma™ Supelco™

Catalog No. 111018797
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018797 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018797 Supplier Merck Emd Millipore Supplier No. 83253100MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 7637-07-2
Quantity 100 mL
MDL Number MFCD00011316
Synonym BF3 - Butanol solution
Grade GC Derivatization
Density 0.87 g/mL (at 20°C)

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany