missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ Boron trichloride Solution, 12% in Methanol, For GC Derivatization, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 10294-34-5 |
| Quantity | 10 x 1 mL |
| Linear Formula | BCl3 |
| MDL Number | MFCD00011313 |
| Formula Weight | 117.17 |
| Grade | GC Derivatization |
| Product Line | LiChropur |
| Density | 0.875-0.895 g/mL (at 20°C) |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany