missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Ammonium formate, Eluent additive for LC-MS, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111014543
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014543 25 g
111014544 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014543 Supplier Merck Emd Millipore Supplier No. 7022125GF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ammonium formate is the ammonium salt of formic acid. It is readily available, is stable and nontoxic in nature. It can be used in conjugation with Palladium on carbon (Pd-C) or Raney-Nickel catalysts.

Chemical Identifiers

MDL Number MFCD00013103
Synonym Formic acid ammonium salt

Specifications

Melting Point 119°C to 121°C
Density 1.26 g/mL (at 25°C)
CAS 540-69-2
pH 5.5 to 7.5 (25°C, 1 M aq. soln.)
Linear Formula HCO2 NH4
Quantity 25 g
Solubility Information Water: 1 M (at 20°C, clear, colorless)
Formula Weight 63.06
Percent Purity ≥99.0% (calc. on dry substance, NT); ≥99.0%
Grade Eluent Additive for LC-MS
Product Line LiChropur
Show More Show Less
RTECSNumber BQ6650000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany