missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ 1,1,1,3,3,3-Hexafluoro-2-Propanol, MilliporeSigma™ Supelco™

Catalog No. 111022432
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022432 10 mL
111022433 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111022432 Supplier Merck Emd Millipore Supplier No. 1812710ML

Chemical Identifiers

CAS 920-66-1
Molecular Formula C3H2F6O
Molecular Weight (g/mol) 168.04
MDL Number MFCD00011651
InChI Key BYEAHWXPCBROCE-UHFFFAOYSA-N
Synonym HFP; Hexafluoroisopropanol
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-ol
SMILES OC(C(F)(F)F)C(F)(F)F

Specifications

Melting Point -4°C (literature)
Density 1.596 g/mL (at 25°C (literature))
Boiling Point 59°C (literature)
Flash Point Closed Cup (Boils Before Flash)
Refractive Index n20/D 1.275 (literature)
Linear Formula (CF3)2 CHOH
Quantity 10 mL
Formula Weight 168.04
Percent Purity ≥99.8% (GC)
Hazard Category H314 - H361fd - H373
RTECSNumber UB6450000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany