missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ 1,1,1,3,3,3-Hexafluoro-2-propanol, For GC derivatization, ≥99.8%, MilliporeSigma™ Supelco™

Catalog No. p-7187757
Change view
Click to view available options
Quantity:
10 mL
50 mL
10 x 1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018364 10 x 1 mL
111018566 10 mL
111017984 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111018364 Supplier Merck Emd Millipore Supplier No. 5251710X1ML
Only null left

Hexafluoroisopropanol (HFP) is generally used to denature the native state of proteins, and it also stabilizes the a-helical conformation in unfolded peptides and proteins like ß-lactoglobulin and melittin.

Chemical Identifiers

MDL Number MFCD00011651
Synonym HFP; Hexafluoroisopropanol

Specifications

Melting Point -4°C
CAS 920-66-1
Density 1.596 g/mL (at 25°C)
Boiling Point 59°C
Linear Formula (CF3)2 CHOH
Quantity 10 x 1 mL
Formula Weight 168.04
Percent Purity ≥99.8% (GC); ≥99.8%
Grade For GC derivatization
Product Line LiChropur
RTECSNumber UB6450000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany