Learn More
3M™ Lexa™ Replacement Lenses
Lexa protective eyewear replacement lens features an 3M™ DX™ Anti-Fog hard coat which helps protect lens from fog, scratching, and many chemicals. Polycarbonate lens absorbs 99.9% UV.
Supplier: 3M™ 152460000020
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Description
Lexa protective eyewear replacement lens features an 3M™ DX™ Anti-Fog hard coat which helps protect lens from fog, scratching, and many chemicals. Polycarbonate lens absorbs 99.9% UV.- Gray, anti-fog lens
- Large size
- 20 safety glasses per case
Specifications
| Anti-fog | |
| Gray | |
| Polycarbonate |
| Three lens angles, and four temple lengths | |
| Replacement Lens |