Learn More
3M™ Lexa™ Replacement Lenses
Catalog No. 19034451
Change view
Click to view available options
Lens Coating:
Anti-fog
Lens Material:
Polycarbonate
Lens Tint:
Clear
Gray
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating Lens Material Lens Tint
19034451 Anti-fog Polycarbonate Clear
19034452 Anti-fog Polycarbonate Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Catalog No. 19034451 Supplier 3M™ Supplier No. 152450000020
Only null left
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Lexa protective eyewear replacement lens features an 3M™ DX™ Anti-Fog hard coat which helps protect lens from fog, scratching, and many chemicals. Polycarbonate lens absorbs 99.9% UV.
Lexa protective eyewear replacement lens features an 3M™ DX™ Anti-Fog hard coat which helps protect lens from fog, scratching, and many chemicals. Polycarbonate lens absorbs 99.9% UV.
  • Gray, anti-fog lens
  • Large size
  • 20 safety glasses per case
Specifications
Lens Coating Anti-fog
Description Three lens angles, and four temple lengths
Lens Tint Clear
Product Type Replacement Lens
Lens Material Polycarbonate