missing translation for 'onlineSavingsMsg'
Learn More

Levofloxacin, 98%

Catalog No. AAJ6694306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6694306 5 g
AAJ6694303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6694306 Supplier Thermo Scientific Chemicals Supplier No. J6694306

Broad-spectrum antibiotic used in pharmacokinetic , antibiotic resistance, and resistance prevention studies. Levofloxacin is active against Gram-positive and Gram-negative bacteria. It inhibits DNA gyrase (type II topoisomerase) and topoisomerase IV, thereby inhibiting cell division. | CAS: 100986-85-4 | C18H20FN3O4 | 361.373 g/mol

Levofloxacin is a broad-spectrum antibiotic used in pharmacokinetic , antibiotic resistance , and resistance prevention studies. Levofloxacin is active against Gram-positive and Gram-negative bacteria. It inhibits DNA gyrase (type II topoisomerase) and topoisomerase IV, thereby inhibiting cell division.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Levofloxacin is a broad-spectrum antibiotic used in pharmacokinetic , antibiotic resistance , and resistance prevention studies. Levofloxacin is active against Gram-positive and Gram-negative bacteria. It inhibits DNA gyrase (type II topoisomerase) and topoisomerase IV, thereby inhibiting cell division.

Solubility
Slightly soluble in water or methanol. Soluble in glacial acetic acid or dichloromethane

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Chemical Identifiers

CAS 100986-85-4
Molecular Formula C18H20FN3O4
Molecular Weight (g/mol) 361.373
MDL Number MFCD00865049
InChI Key GSDSWSVVBLHKDQ-JTQLQIEISA-N
PubChem CID 149096
ChEBI CHEBI:63598
SMILES CC1COC2=C3N1C=C(C(=O)C3=CC(=C2N4CCN(CC4)C)F)C(=O)O

Specifications

Type Levofloxacin
Physical Form Powder
Melting Point 218°C to 227°C
Packaging Glass Bottle
Quantity 5 g
Flash Point >110°C (230°F)
Sensitivity Light sensitive
Boiling Point 572°C
Beilstein 7327015
Solubility Information Slightly soluble in water or methanol. Soluble in glacial acetic acid or dichloromethane
Formula Weight 361.37
Percent Purity 98%
Chemical Name or Material Levofloxacin
Show More Show Less
Hazard Category H302-H317-H334
Precautionary Statement P261-P264b-P270-P272-P280g-P285-P301+P312-P302+P352-P304+P341-P330-P333+P313-P342+P311-P363-P501c
RTECSNumber UU8815550
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only