missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Levocarnitine, Free Base, USP, 97-103%, Spectrum™ Chemical
C1399, 541-15-1, C7H15NO3
Supplier: Spectrum Chemical Mfg Cor C13995KGBL
Description
Spectrum™ Chemical Levocarnitine, Free Base, USP is used to prevent and treat a lack of carnitine. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 541-15-1 | |
| C7H15NO3 | |
| PHIQHXFUZVPYII-UHFFFAOYNA-N | |
| 3-hydroxy-4-(trimethylazaniumyl)butanoate | |
| 97 to 103% | |
| 0.5% |
| 100% | |
| 5 kg | |
| C[N+](C)(C)CC(O)CC([O-])=O | |
| 161.20 | |
| USP | |
| Poly Pail |
Chemical Identifiers
| 541-15-1 | |
| 161.20 | |
| 3-hydroxy-4-(trimethylazaniumyl)butanoate |
| C7H15NO3 | |
| PHIQHXFUZVPYII-UHFFFAOYNA-N | |
| C[N+](C)(C)CC(O)CC([O-])=O |