missing translation for 'onlineSavingsMsg'
Learn More

Levetiracetam 98.0+%, TCI America™

Catalog No. L02345G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L02345G 5 g
L02341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L02345G Supplier TCI America Supplier No. L02345G

Chemical Identifiers

CAS 102767-28-2
Molecular Formula C8H14N2O2
Molecular Weight (g/mol) 170.21
MDL Number MFCD03265610
InChI Key HPHUVLMMVZITSG-ZCFIWIBFSA-N
Synonym levetiracetam, keppra, s-2-2-oxopyrrolidin-1-yl butanamide, keppra xr, 2s-2-2-oxopyrrolidin-1-yl butanamide, levetiracetamum, ucb-l 059, ucb l059, levetiractam, levroxa
PubChem CID 5284583
ChEBI CHEBI:6437
IUPAC Name (2R)-2-(2-oxopyrrolidin-1-yl)butanamide
SMILES CC[C@@H](N1CCCC1=O)C(N)=O

Specifications

Melting Point 118°C
Color White
Quantity 5 g
Formula Weight 170.21
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Levetiracetam
RTECSNumber UX9656166
TSCA No