missing translation for 'onlineSavingsMsg'
Learn More

High-Tech Conversions Bee-Safe™ Isolation Gowns

Catalog No. 17444084
Change view
Click to view available options
Size:
Large
X-Large
Color:
Blue
Style:
Level 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color Style
17444084 X-Large Blue Level 2
17444083 Large Blue Level 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17444084 Supplier High-Tech Conversions Supplier No. GAHGISO2BX

Spunbond polypropylene isolation gowns are fluid resistant and ideal for a variety of healthcare settings. Choose from AAMI Level 1 or Level 2 styles.

  • TESTING: Level 1 Gown
    • Liquid Barrier: ANSI/AAMI PB70
    • Water Resistance (Impact Penetration Test): AATCC 42
    • Water Resistance (Hydrostatic Pressure Test): AATCC 127
  • TESTING: Level 2 Gown
    • Liquid Barrier: ANSI/AAMI PB70
    • Water Resistance (Impact Penetration Test): AATCC 42
    • Water Resistance (Hydrostatic Pressure Test): AATCC 127

Specifications

Style Level 2
Gender Unisex
Product Type Isolation Gown
No. of Pockets None
For Use With (Application) Healthcare Settings
Size X-Large
Material Laminated Spunbond Polypropylene
Color Blue
Disposable Single-use
Sterility Non-sterile
Cuff Style Knit
Latex Free Yes
Model GAH-GISO2BX
Sleeve Style Long Sleeve
Certifications/Compliance ANSI/AAMI PB70, AATCC 42, AATCC 127
Autoclavable Not Autoclavable
No. per Pack 10/Bag, 5 Bags/Cs.
Show More Show Less