missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Leuprolide

For cellular and molecular biology applications

$347.77 - $347.77

Specifications

Purification Method HPLC
Common Name Leuprorelin
Format Powder
For Use With (Application) Highly active luteinizing hormone releasing hormone (LHRH) agonist
Molecular Weight (g/mol) 1209.41
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62147LB0
View Documents
Thermo Scientific Chemicals
J62147LB0
5 mg
Each for $347.77
Only null left
 
Description

Description

Highly active luteinizing hormone releasing hormone (LHRH) agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

HPLC
Powder
1209.41
RUO - research use only
Leuprorelin
Highly active luteinizing hormone releasing hormone (LHRH) agonist
Leuprolide
[Des-Gly10, D-Leu6, Pro-NHEt9]LH-RH
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only