Learn More
Greiner Bio-One Leucosep Centrifuge Tubes with Porous Barrier
GREENER_CHOICE
Efficient separation of lymphocytes and mononuclear cells.
Supplier:  Greiner Bio-One 227290P
 View more versions of this product
Catalog No. 07000983
Only null left
Description
Description
  • Non-cytotoxic, non-pyrogenic
  • Enrichment directly from whole blood
  • Simplified filling through porous barrier
  • Rapid separation in 15 minutes at room temperature
  • No additional laboratory equipment required, asepticall
    Service Plan
    No
Specifications
Chemical Identifiers
Natural
Polypropylene
Yes
Centrifuge Tube
Translucent; print of graduation: blue
Sterile
Yes
Specifications
Natural
Polypropylene
115 mm
Yes
Centrifuge Tube
Translucent; print of graduation: blue
Sterile
Conical
Yes
Product Suggestions
Product Suggestions